2,3-Dibromo-5-nitrotoluene ≥97%

Supplier: AOBChem USA

Warning

87650-5G 87650-25G 87650-1G 87650-500MG
77635-344EA 431 USD
77635-344 77657-004 77644-156 77652-134
2,3-Dibromo-5-nitrotoluene ≥97%
2,3-Dibromo-5-nitrotoluene

Precaución: For Research Use Only

Formula: C₇H₅Br₂NO₂
MW: 294.928 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846021
CAS Number: 73557-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR