7-Methyl-2,3-dihydro-1,4-benzodioxine-6-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

87635-5G 87635-1G 87635-500MG 87635-25G
77635-342EA 442 USD
77635-342 77640-128 77653-116 77657-002
7-Methyl-2,3-dihydro-1,4-benzodioxine-6-carbaldehyde ≥95%
7-Methyl-2,3-dihydro-1,4-benzodioxine-6-carbaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD11053948
CAS Number: 724791-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR