2-Bromo-3-fluoro-6-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

87305-5G 87305-1G 87305-500MG
77660-470EA 1802 USD
77660-470 77644-150 77648-124
2-Bromo-3-fluoro-6-methoxyphenol ≥97%
2-Bromo-3-fluoro-6-methoxyphenol

Precaución: For Research Use Only

Formula: C₇H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28738843
CAS Number: 1780209-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR