(3-Methoxy-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

86423-5G 86423-25G 86423-1G 86423-500MG
77635-338EA 288 USD
77635-338 77656-994 77644-146 77648-120
(3-Methoxy-5-methylphenyl)(methyl)sulfane ≥97%
(3-Methoxy-5-methylphenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: 168.256 g/mol
MW: C₉H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD22489308
CAS Number: 66623-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR