Trimethyl(2-methyl-5-(trifluoromethyl)phenyl)silane ≥95%

Supplier: AOBChem USA

Warning

86349-5G 86349-1G 86349-500MG
77630-792EA 1658 USD
77630-792 77648-116 77648-118
Trimethyl(2-methyl-5-(trifluoromethyl)phenyl)silane ≥95%
Trimethyl(2-methyl-5-(trifluoromethyl)phenyl)silane

Precaución: For Research Use Only

Formula: 232.318 g/mol
MW: C₁₁H₁₅F₃Si
Storage Temperature: Ambient
MDL Number: MFCD34687740
CAS Number: 2153271-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR