(4-Chloro-2-fluoro-3-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

86278-5G 86278-1G 86278-500MG 86278-25G
77633-814EA 868 USD
77633-814 77640-120 77653-112 77660-466
(4-Chloro-2-fluoro-3-methylphenyl)trimethylsilane ≥95%
(4-Chloro-2-fluoro-3-methylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: C₁₀H₁₄ClFSi
Storage Temperature: Ambient
MDL Number: MFCD30303982
CAS Number: 1864342-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR