(5-Fluoro-2-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

86153-5G 86153-25G 86153-1G 86153-500MG
77633-812EA 890 USD
77633-812 77660-460 77644-144 77652-124
(5-Fluoro-2-methylphenyl)trimethylsilane ≥95%
(5-Fluoro-2-methylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: C₁₀H₁₅FSi
Storage Temperature: Ambient
MDL Number: MFCD11521276
CAS Number: 1314911-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR