(4-Fluoro-3,5-dimethylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

86139-5G 86139-1G 86139-25G 86139-500MG
77633-810EA 1180 USD
77633-810 77640-114 77660-458 77652-122
(4-Fluoro-3,5-dimethylphenyl)trimethylsilane ≥95%
(4-Fluoro-3,5-dimethylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: C₁₁H₁₇FSi
Storage Temperature: Ambient
MDL Number: MFCD23143148
CAS Number: 1443311-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR