(3-Isopropylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

86125-5G 86125-1G 86125-25G 86125-500MG
77633-806EA 1180 USD
77633-806 77640-110 77660-456 77652-120
(3-Isopropylphenyl)trimethylsilane ≥95%
(3-Isopropylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: C₁₂H₂₀Si
Storage Temperature: Ambient
MDL Number: MFCD11521275
CAS Number: 18027-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR