1-(3-(Trifluoromethyl)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

86010-5G 86010-1G 86010-25G 86010-500MG
77630-790EA 1441 USD
77630-790 77640-108 77660-454 77652-118
1-(3-(Trifluoromethyl)phenyl)cyclopentanol ≥95%
1-(3-(Trifluoromethyl)phenyl)cyclopentanol

Precaución: For Research Use Only

Formula: C₁₂H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD17222248
CAS Number: 1342610-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR