1-(4-Chloro-2-fluoro-3-methylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85988-5G 85988-1G 85988-500MG
77630-788EA 1688 USD
77630-788 77640-104 77648-112
1-(4-Chloro-2-fluoro-3-methylphenyl)cyclopentanol ≥95%
1-(4-Chloro-2-fluoro-3-methylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 228.69 g/mol
MW: C₁₂H₁₄ClFO
Storage Temperature: Ambient
MDL Number: MFCD34770596
CAS Number: 1528210-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR