1-(3-(Benzyloxy)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85951-5G 85951-1G 85951-25G 85951-500MG
77633-800EA 1381 USD
77633-800 77640-102 77656-986 77648-110
1-(3-(Benzyloxy)phenyl)cyclopentanol ≥95%
1-(3-(Benzyloxy)phenyl)cyclopentanol

Precaución: For Research Use Only

Formula: C₁₈H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD34770572
CAS Number: 211495-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR