1-(4-Methoxy-2-methylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85945-5G 85945-1G 85945-25G 85945-500MG
77633-798EA 1381 USD
77633-798 77640-100 77656-984 77648-108
1-(4-Methoxy-2-methylphenyl)cyclopentanol ≥95%
1-(4-Methoxy-2-methylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD21216089
CAS Number: 1484650-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR