1-(3-Fluoro-4-methylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85926-5G 85926-25G 85926-1G 85926-500MG
77630-786EA 1441 USD
77630-786 77660-452 77644-140 77652-112
1-(3-Fluoro-4-methylphenyl)cyclopentanol ≥95%
1-(3-Fluoro-4-methylphenyl)cyclopentanol
Formula: C₁₂H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD34770564
CAS Number: 1702717-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR