1-(2,5-Dimethylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85885-5G 85885-1G 85885-25G 85885-500MG
77630-782EA 1441 USD
77630-782 77640-096 77660-448 77652-110
1-(2,5-Dimethylphenyl)cyclopentanol ≥95%
1-(2,5-Dimethylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD16794463
CAS Number: 1250368-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR