1-(3,4-Dimethylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85884-5G 85884-1G 85884-25G 85884-500MG
77630-780EA 1441 USD
77630-780 77640-094 77656-980 77648-100
1-(3,4-Dimethylphenyl)cyclopentanol ≥95%
1-(3,4-Dimethylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 190.281 g/mol
MW: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD00559294
CAS Number: 1250573-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR