1-(4-Propylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85883-5G 85883-25G 85883-1G 85883-500MG
77633-794EA 1206 USD
77633-794 77660-446 77644-138 77652-108
1-(4-Propylphenyl)cyclopentanol ≥95%
1-(4-Propylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 204.308 g/mol
MW: C₁₄H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD17223764
CAS Number: 1342662-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR