1-(3,5-Dimethylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85872-5G 85872-25G 85872-1G 85872-500MG
77630-778EA 1441 USD
77630-778 77660-444 77644-136 77648-098
1-(3,5-Dimethylphenyl)cyclopentanol ≥95%
1-(3,5-Dimethylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 190.281 g/mol
MW: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD12806370
CAS Number: 1179185-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR