1-(2-Fluorophenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85840-5G 85840-25G 85840-1G 85840-500MG
77633-792EA 998 USD
77633-792 77660-442 77644-132 77652-104
1-(2-Fluorophenyl)cyclopentanol ≥95%
1-(2-Fluorophenyl)cyclopentanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD12152935
CAS Number: 1250703-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR