1-(4-Fluorophenyl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

85836-5G 85836-1G 85836-500MG
77630-772EA 1675 USD
77630-772 77644-130 77648-094
1-(4-Fluorophenyl)cyclopentanol ≥97%
1-(4-Fluorophenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 180.219 g/mol
MW: C₁₁H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD00045410
CAS Number: 339366-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR