1-(2-(Benzyloxy)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85821-5G 85821-1G 85821-25G 85821-500MG
77630-770EA 1441 USD
77630-770 77640-090 77656-978 77652-100
1-(2-(Benzyloxy)phenyl)cyclobutanol ≥95%
1-(2-(Benzyloxy)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₇H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34770543
CAS Number: 1402004-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR