1-(2,3-Dichlorophenyl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

85818-5G 85818-25G 85818-1G 85818-500MG
77630-768EA 1488 USD
77630-768 77636-232 77644-126 77652-098
1-(2,3-Dichlorophenyl)cyclobutanol ≥97%
1-(2,3-Dichlorophenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 217.092 g/mol
MW: C₁₀H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD21788596
CAS Number: 1512242-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR