1-(3-Chloro-4-methoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85816-5G 85816-25G 85816-1G 85816-500MG
77630-766EA 1441 USD
77630-766 77660-438 77644-124 77648-092
1-(3-Chloro-4-methoxyphenyl)cyclobutanol ≥95%
1-(3-Chloro-4-methoxyphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34770541
CAS Number: 1516225-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR