1-(2,5-Dichlorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85815-5G 85815-1G 85815-500MG
77630-764EA 1553 USD
77630-764 77644-122 77648-090
1-(2,5-Dichlorophenyl)cyclobutanol ≥95%
1-(2,5-Dichlorophenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 217.092 g/mol
MW: C₁₀H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD21788592
CAS Number: 1519530-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR