1-(2,5-Dimethylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85814-5G 85814-25G 85814-1G 85814-500MG
77630-762EA 1488 USD
77630-762 77636-230 77644-120 77648-088
1-(2,5-Dimethylphenyl)cyclobutanol ≥95%
1-(2,5-Dimethylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 176.255 g/mol
MW: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD21788625
CAS Number: 1519546-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR