1-(3-Fluoro-2-methoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85812-5G 85812-25G 85812-1G 85812-500MG
77630-760EA 1441 USD
77630-760 77660-436 77644-118 77648-086
1-(3-Fluoro-2-methoxyphenyl)cyclobutanol ≥95%
1-(3-Fluoro-2-methoxyphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34770540
CAS Number: 1621475-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR