1-(3-Fluoro-4-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85810-5G 85810-1G 85810-500MG
77630-758EA 1553 USD
77630-758 77640-088 77652-096
1-(3-Fluoro-4-methylphenyl)cyclobutanol ≥95%
1-(3-Fluoro-4-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 180.219 g/mol
MW: C₁₁H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD30164393
CAS Number: 1702076-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR