1-(3,5-Bis(trifluoromethyl)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85803-5G 85803-1G 85803-500MG 85803-25G
77633-790EA 1288 USD
77633-790 77640-086 77653-108 77660-434
1-(3,5-Bis(trifluoromethyl)phenyl)cyclobutanol ≥95%
1-(3,5-Bis(trifluoromethyl)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 284.198 g/mol
MW: C₁₂H₁₀F₆O
Storage Temperature: Ambient
MDL Number: MFCD34770537
CAS Number: 2242185-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR