1-(4-(Trimethylsilyl)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85802-1G 85802-5G 85802-500MG
77640-084EA 676 USD
77640-084 77660-432 77652-092
1-(4-(Trimethylsilyl)phenyl)cyclobutanol ≥95%
1-(4-(Trimethylsilyl)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₃H₂₀OSi
Storage Temperature: Ambient
MDL Number: MFCD34770536
CAS Number: 2282437-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR