1-(2,6-Difluorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85781-1G 85781-500MG 85781-5G
77640-082EA 751 USD
77640-082 77653-106 77656-974
1-(2,6-Difluorophenyl)cyclobutanol ≥95%
1-(2,6-Difluorophenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₀H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD21788731
CAS Number: 935287-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR