1-(4-Methylnaphthalen-1-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85771-5G 85771-1G 85771-500MG
77630-756EA 1688 USD
77630-756 77640-080 77653-104
1-(4-Methylnaphthalen-1-yl)cyclobutanol ≥95%
1-(4-Methylnaphthalen-1-yl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₅H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD34770520
CAS Number: 2022068-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR