1-(2,4-Dichlorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85757-5G 85757-1G 85757-500MG
77656-972EA 1898 USD
77656-972 77644-114 77648-084
1-(2,4-Dichlorophenyl)cyclobutanol ≥95%
1-(2,4-Dichlorophenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₀H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD29987427
CAS Number: 1540838-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR