1-(4-Phenoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85730-5G 85730-25G 85730-1G 85730-500MG
77630-752EA 1441 USD
77630-752 77660-428 77644-112 77648-082
1-(4-Phenoxyphenyl)cyclobutanol ≥95%
1-(4-Phenoxyphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 240.297 g/mol
MW: C₁₆H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34770508
CAS Number: 1946772-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR