1-(3,5-Dimethylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85725-5G 85725-1G 85725-500MG
77630-750EA 1688 USD
77630-750 77644-110 77648-080
1-(3,5-Dimethylphenyl)cyclobutanol ≥95%
1-(3,5-Dimethylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 176.255 g/mol
MW: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD21788799
CAS Number: 1501510-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR