1-(Naphthalen-1-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85721-5G 85721-25G 85721-1G 85721-500MG
77633-788EA 1357 USD
77633-788 77660-426 77648-076 77648-078
1-(Naphthalen-1-yl)cyclobutanol ≥95%
1-(Naphthalen-1-yl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₄H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD21788763
CAS Number: 74685-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR