1-([1,1'-Biphenyl]-4-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85719-5G 85719-1G 85719-500MG
77630-748EA 1553 USD
77630-748 77640-074 77653-102
1-([1,1'-Biphenyl]-4-yl)cyclobutanol ≥95%
1-([1,1'-Biphenyl]-4-yl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₆H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD22235050
CAS Number: 1500032-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR