1-(Naphthalen-2-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85717-5G 85717-25G 85717-1G 85717-500MG
77633-784EA 1357 USD
77633-784 77656-970 77644-108 77652-086
1-(Naphthalen-2-yl)cyclobutanol ≥95%
1-(Naphthalen-2-yl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₄H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD21788773
CAS Number: 57350-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR