4'-Iodo-3-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

85423-5G 85423-1G 85423-500MG
77630-746EA 1788 USD
77630-746 77644-106 77648-072
4'-Iodo-3-methyl-1,1'-biphenyl ≥95%
4'-Iodo-3-methyl-1,1'-biphenyl

Precaución: For Research Use Only

Formula: C₁₃H₁₁I
Storage Temperature: Ambient
MDL Number: MFCD22494917
CAS Number: 1312304-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR