2-Chloro-4-formylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

85365-5G 85365-500MG 85365-25G 85365-1G
77633-782EA 719 USD
77633-782 77653-100 77660-422 77644-104
2-Chloro-4-formylbenzonitrile ≥95%
2-Chloro-4-formylbenzonitrile

Precaución: For Research Use Only

Formula: 165.576 g/mol
MW: C₈H₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD18392411
CAS Number: 101048-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR