2-Bromo-6-chloro-3-ethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

85247-5G 85247-25G 85247-1G 85247-500MG
77630-742EA 1441 USD
77630-742 77660-418 77644-100 77652-084
2-Bromo-6-chloro-3-ethoxybenzaldehyde ≥95%
2-Bromo-6-chloro-3-ethoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₈BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD34817260
CAS Number: 2385298-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR