1-(4-Bromo-3-ethylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

84664-5G 84664-1G 84664-500MG
77656-966EA 2417 USD
77656-966 77644-098 77652-082
1-(4-Bromo-3-ethylphenyl)ethanol ≥97%
1-(4-Bromo-3-ethylphenyl)ethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD34770112
CAS Number: 2918924-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR