(3-Bromo-2,6-difluorophenyl)(piperidin-1-yl)methanone ≥95%
Supplier: AOBChem USA
|
Warning
|
84545-5G
84545-1G
84545-25G
84545-500MG
77633-778EA
898
USD
77633-778
77640-070
77660-416
77648-066
(3-Bromo-2,6-difluorophenyl)(piperidin-1-yl)methanone ≥95%
(3-Bromo-2,6-difluorophenyl)(piperidin-1-yl)methanone
Precaución: For Research Use Only
Formula:
C₁₂H₁₂BrF₂NO Storage Temperature: Ambient |
MDL Number:
MFCD34770071 CAS Number: 2918914-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...