2-Bromo-1-chloro-5-iodo-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

83987-5G 83987-1G 83987-500MG
77630-736EA 1788 USD
77630-736 77640-068 77652-080
2-Bromo-1-chloro-5-iodo-3-methylbenzene ≥95%
2-Bromo-1-chloro-5-iodo-3-methylbenzene

Precaución: For Research Use Only

Formula: 331.376 g/mol
MW: C₇H₅BrClI
Storage Temperature: Ambient
MDL Number: MFCD20233420
CAS Number: 1349718-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR