4-(Trimethylsilyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

83956-5G 83956-1G 83956-25G 83956-500MG
77635-334EA 417 USD
77635-334 77640-066 77656-964 77648-062
4-(Trimethylsilyl)benzaldehyde ≥95%
4-(Trimethylsilyl)benzaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₁₄OSi
Storage Temperature: Ambient
MDL Number: MFCD08064053
CAS Number: 2199-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR