Tert-butyl 2-bromo-6-fluoro-4-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

83943-5G 83943-25G 83943-1G 83943-500MG
77630-734EA 1441 USD
77630-734 77636-226 77640-064 77648-060
Tert-butyl 2-bromo-6-fluoro-4-(trifluoromethyl)benzoate ≥95%
Tert-butyl 2-bromo-6-fluoro-4-(trifluoromethyl)benzoate

Precaución: For Research Use Only

Formula: C₁₂H₁₁BrF₄O₂
MW: 343.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769729
CAS Number: 2451903-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR