Tert-butyl 2-fluoro-5-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

83762-5G 83762-1G 83762-500MG
77630-732EA 1610 USD
77630-732 77644-094 77652-078
Tert-butyl 2-fluoro-5-methylbenzoate ≥95%
Tert-butyl 2-fluoro-5-methylbenzoate

Precaución: For Research Use Only

Formula: 210.245 g/mol
MW: C₁₂H₁₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD18897598
CAS Number: 1343831-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR