4-(2-Bromophenoxy)-2-methylaniline ≥95%

Supplier: AOBChem USA

Warning

83607-5G 83607-1G 83607-25G 83607-500MG
77630-730EA 1441 USD
77630-730 77640-060 77656-960 77652-076
4-(2-Bromophenoxy)-2-methylaniline ≥95%
4-(2-Bromophenoxy)-2-methylaniline

Precaución: For Research Use Only

Formula: 278.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12169255
CAS Number: 1154154-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR