2-Ethoxy-1-ethyl-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

83449-5G 83449-25G 83449-1G 83449-500MG
77630-728EA 1441 USD
77630-728 77660-412 77644-092 77652-074
2-Ethoxy-1-ethyl-3-iodobenzene ≥97%
2-Ethoxy-1-ethyl-3-iodobenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₃IO
Storage Temperature: Ambient
MDL Number: MFCD34769524
CAS Number: 2383076-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR