(2-Ethoxy-3-ethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

83448-5G 83448-500MG 83448-25G 83448-1G
77633-774EA 1441 USD
77633-774 77653-098 77656-958 77648-056
(2-Ethoxy-3-ethylphenyl)(methyl)sulfane ≥97%
(2-Ethoxy-3-ethylphenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: C₁₁H₁₆OS
Storage Temperature: Ambient
MDL Number: MFCD34769523
CAS Number: 2386970-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR