2,2-Dichloro-1-(2,5-difluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

83079-5G 83079-1G 83079-500MG
77656-956EA 2348 USD
77656-956 77644-090 77652-072
2,2-Dichloro-1-(2,5-difluorophenyl)ethanone ≥95%
2,2-Dichloro-1-(2,5-difluorophenyl)ethanone

Precaución: For Research Use Only

Formula: 225.02 g/mol
MW: C₈H₄Cl₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD28105919
CAS Number: 1499796-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR